Order Entry
Finland
ContactUsLinkComponent
3-(4-Fluoro-phenylsulphanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
3-(4-Fluoro-phenylsulphanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408331-500MG
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Fluoro-phenylsulphanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408331-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :325.441 g/mol
Storage Temperature:Ambient
CAS Number:1289388-37-9
Formula:C₁₇H₂₄FNO₂S
MDL Number:MFCD18837557