
6-Fluoro-1H-indole-4-boronic acid, pinacol ester
Catalog # APOSPC400528-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:261.1 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:955978-85-5
Formula:C₁₄H₁₇BFNO₂
MDL Number:MFCD16987817