Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4’-Bromo-3’-chloro-2’-fluorophenacyl bromide
4’-Bromo-3’-chloro-2’-fluorophenacyl bromide
4’-Bromo-3’-chloro-2’-fluorophenacyl bromide
4’-Bromo-3’-chloro-2’-fluorophenacyl bromide
Supplier:
Apollo Scientific
Certificates
Expand image
4’-Bromo-3’-chloro-2’-fluorophenacyl bromide
Supplier:
Apollo Scientific
About this item
CAS Number:
1805582-66-4
:
Formula:
C₈H₄Br₂ClFO
MDL Number:
MFCD28733994
:
330.376 g/mol
Storage Temperature:
Ambient
UN:
3261
ADR:
8,III
:
GHS05