Pentafluorophenyl piperidine-4-carboxylate, N-BOC protected
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:294885-28-2
:
Formula:C₁₇H₁₈F₅NO₄
MDL Number:MFCD09879992
:395.321 g/mol
Melting Pt:84…86.5 °C
Storage Temperature:Ambient