0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
3-Bromo-5-chloro-2,6-difluorobenzyl bromide
3-Bromo-5-chloro-2,6-difluorobenzyl bromide 97%
3-Bromo-5-chloro-2,6-difluorobenzyl bromide 97%
3-Bromo-5-chloro-2,6-difluorobenzyl bromide 97%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Bromo-5-chloro-2,6-difluorobenzyl bromide 97%
Supplier:
Apollo Scientific
About this item
:
Formula:
C₇H₃Br₂ClF₂
MDL Number:
MFCD32640106
:
320.357 g/mol
Storage Temperature:
Ambient