Order Entry
Finland
ContactUsLinkComponent
5-Chloro-2,4-difluorophenylacetonitrile 97%
5-Chloro-2,4-difluorophenylacetonitrile 97%
Catalog # APOSPC303356-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-2,4-difluorophenylacetonitrile 97%
Catalog # APOSPC303356-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :187.574 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
CAS Number:1429422-26-3
Formula:C₈H₄ClF₂N
MDL Number:MFCD28402683