Order Entry
Finland
ContactUsLinkComponent
1-[2-Methyl-3-(trifluoromethyl)phenyl]-1H-pyrrole-2-carbaldehyde
1-[2-Methyl-3-(trifluoromethyl)phenyl]-1H-pyrrole-2-carbaldehyde
Catalog # APOSPC300919-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[2-Methyl-3-(trifluoromethyl)phenyl]-1H-pyrrole-2-carbaldehyde
Catalog # APOSPC300919-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :253.22 g/mol
Storage Temperature:Ambient
CAS Number:2197063-05-9
Formula:C₁₃H₁₀F₃NO
MDL Number:MFCD30481160