2-Methyl-6-(trifluoromethyl)-2H-1,4-benzoxazin-3(4H)-one 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:189940-07-6
:
Formula:C₁₀H₈F₃NO₂
MDL Number:MFCD19376607
:231.171 g/mol
Melting Pt:154…156 °C
Storage Temperature:Ambient