Order Entry
Finland
ContactUsLinkComponent
1-(2-Fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid, tech
1-(2-Fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid, tech
Catalog # APOSPC28082-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid, tech
Catalog # APOSPC28082-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :234.226 g/mol
Storage Temperature:Ambient
CAS Number:956930-31-7
Formula:C₁₂H₁₁FN₂O₂
MDL Number:MFCD07021455