
Perfluoro(1,2-dichlorocyclobut-1-ene) 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:377-93-5
:
Formula:C₄Cl₂F₄
MDL Number:MFCD00019268
Boiling Pt:67.1 °C (760 mm Hg)
Melting Pt:–43 °C
Density:1.534 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:194.942 g/mol
:GHS07
Frequently Bought Together