

(2-Bromo-1,1-difluoroethyl)benzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:108661-89-8
:
Formula:C₈H₇BrF₂
MDL Number:MFCD07777119
:221.042 g/mol
Boiling Pt:55…56 °C (2 mm Hg)
Melting Pt:–26…–24 °C
Density:1.534 g/cm³ (25 °C)
:GHS05
:GHS07
Storage Temperature:Ambient