Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-5-fluorobenzene-1-sulphonyl chloride
3-Bromo-5-fluorobenzene-1-sulphonyl chloride 95%
3-Bromo-5-fluorobenzene-1-sulphonyl chloride 95%
3-Bromo-5-fluorobenzene-1-sulphonyl chloride 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Bromo-5-fluorobenzene-1-sulphonyl chloride 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1214342-44-5
:
Formula:
C₆H₃BrClFO₂S
MDL Number:
MFCD13185364
:
273.507 g/mol
Storage Temperature:
Ambient