Methyl 6-oxo-1-{[4-(trifluoromethyl)phenyl]methyl}-1,6-dihydropyridine-3-carboxylate
Catalog # APOSPC200453-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:311.256 g/mol
Melting Pt:103…105 °C
Storage Temperature:Ambient
CAS Number:1582770-08-8
Formula:C₁₅H₁₂F₃NO₃
MDL Number:MFCD27980590