1-(1-{[3-(Trifluoromethyl)phenyl]methyl}-1H-1,2,3-triazol-4-yl)ethan-1-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1443291-23-3
:
Formula:C₁₂H₁₀F₃N₃O
MDL Number:MFCD25368740
:269.223 g/mol
Melting Pt:105…107 °C
Storage Temperature:Ambient