Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Amino-2-(trifluoromethyl)benzonitrile
3-Amino-2-(trifluoromethyl)benzonitrile
3-Amino-2-(trifluoromethyl)benzonitrile
3-Amino-2-(trifluoromethyl)benzonitrile
Supplier:
Apollo Scientific
Certificates
Expand image
3-Amino-2-(trifluoromethyl)benzonitrile
Supplier:
Apollo Scientific
About this item
CAS Number:
1369869-39-5
:
Formula:
C₈H₅F₃N₂
MDL Number:
MFCD22480978
:
186.134 g/mol
Storage Temperature:
Ambient
UN:
3439
ADR:
6.1,III
:
GHS06