Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Methoxy-3,4-dihydro-1H-quinolin-2-one
6-Methoxy-3,4-dihydro-1H-quinolin-2-one 98%
6-Methoxy-3,4-dihydro-1H-quinolin-2-one 98%
Irritant
6-Methoxy-3,4-dihydro-1H-quinolin-2-one 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
6-Methoxy-3,4-dihydro-1H-quinolin-2-one 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
54197-64-7
:
Formula:
C₁₀H₁₁NO₂
MDL Number:
MFCD02179395
:
177.2 g/mol
:
GHS07
Storage Temperature:
Ambient