Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acridine-9-carboxylic acid
Acridine-9-carboxylic acid
Acridine-9-carboxylic acid
Irritant
Acridine-9-carboxylic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Acridine-9-carboxylic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
5336-90-3
:
Formula:
C₁₄H₉NO₂
MDL Number:
MFCD00009734
:
223.227 g/mol
Melting Pt:
290 °C
:
GHS07
Storage Temperature:
Ambient