Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R)-3-Amino-3-(3-chlorophenyl)propionic acid
(R)-3-Amino-3-(3-chlorophenyl)propionic acid 97%
(R)-3-Amino-3-(3-chlorophenyl)propionic acid 97%
(R)-3-Amino-3-(3-chlorophenyl)propionic acid 97%
Supplier:
Apollo Scientific
Certificates
Expand image
(R)-3-Amino-3-(3-chlorophenyl)propionic acid 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
262429-49-2
:
Formula:
C₉H₁₀ClNO₂
MDL Number:
MFCD04113654
:
199.634 g/mol
Storage Temperature:
Ambient