Order Entry
Finland
ContactUsLinkComponent
1-(3-Methylbutyl)-1H-pyrazol-5-ol 95%
1-(3-Methylbutyl)-1H-pyrazol-5-ol 95%
Catalog # APOSOR957445-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(3-Methylbutyl)-1H-pyrazol-5-ol 95%
Catalog # APOSOR957445-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :154.21 g/mol
Storage Temperature:Ambient
Formula:C₈H₁₄N₂O
MDL Number:MFCD26070675