Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-1,5-dimethyl-1H-pyrrole-2-carbonitrile
3-Bromo-1,5-dimethyl-1H-pyrrole-2-carbonitrile 95%
3-Bromo-1,5-dimethyl-1H-pyrrole-2-carbonitrile 95%
3-Bromo-1,5-dimethyl-1H-pyrrole-2-carbonitrile 95%
APOSOR956691-1G
:
Apollo Scientific
:
3-Bromo-1,5-dimethyl-1H-pyrrole-2-carbonitrile 95%
APOSOR956691-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
199.048 g/mol
:
Ambient
:
942612-14-8
:
C₇H₇BrN₂
:
MFCD21336851