Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Ethoxy-2-nitrobenzonitrile
5-Ethoxy-2-nitrobenzonitrile 96%
5-Ethoxy-2-nitrobenzonitrile 96%
5-Ethoxy-2-nitrobenzonitrile 96%
Supplier:
Apollo Scientific
Certificates
Expand image
5-Ethoxy-2-nitrobenzonitrile 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
1415819-87-2
:
Formula:
C₉H₈N₂O₃
MDL Number:
MFCD22586697
:
192.171 g/mol
Storage Temperature:
Ambient