2-Methyl-1-(2-pyridyl)-1-propylamine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:58088-72-5
:
Formula:C₉H₁₄N₂
MDL Number:MFCD09802325
:150.221 g/mol
Storage Temperature:Refrigerator
UN:2810
ADR:6.1,III
Frequently Bought Together