Order Entry
Finland
ContactUsLinkComponent
6-Ethyl-4-methoxy-2-pyranone 97%
6-Ethyl-4-methoxy-2-pyranone 97%
Catalog # APOSOR952337-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Ethyl-4-methoxy-2-pyranone 97%
Catalog # APOSOR952337-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :154.163 g/mol
Storage Temperature:Ambient
CAS Number:106950-13-4
Formula:C₈H₁₀O₃
MDL Number:MFCD09991688