Order Entry
Finland
ContactUsLinkComponent
(1R)-1-(4-Chlorophenyl)-1,2-ethanediol 95%
(1R)-1-(4-Chlorophenyl)-1,2-ethanediol 95%
Catalog # APOSOR947779-1G
Supplier:  Apollo Scientific
CAS Number:  
(1R)-1-(4-Chlorophenyl)-1,2-ethanediol 95%
Catalog # APOSOR947779-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :172.609 g/mol
Storage Temperature:Ambient
CAS Number:152142-03-5
Formula:C₈H₉ClO₂
MDL Number:MFCD19689566