Order Entry
Finland
ContactUsLinkComponent
2-Bromo-6-methylisonicotinic acid ≥98%
2-Bromo-6-methylisonicotinic acid ≥98%
Catalog # APOSOR946687-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-6-methylisonicotinic acid ≥98%
Catalog # APOSOR946687-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :216.032 g/mol
Storage Temperature:Refrigerator
CAS Number:25462-84-4
Formula:C₇H₆BrNO₂
MDL Number:MFCD13188656