Order Entry
Finland
ContactUsLinkComponent
6-Methyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid methyl ester 97%
6-Methyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid methyl ester 97%
Catalog # APOSOR940815-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Methyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid methyl ester 97%
Catalog # APOSOR940815-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.162 g/mol
Storage Temperature:Ambient
CAS Number:51146-06-6
 :MFCD09953605
Formula:C₈H₉NO₃
MDL Number:MFCD00584442