0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
4-Chloroquinolin-8-amine
4-Chloroquinolin-8-amine 95%
4-Chloroquinolin-8-amine 95%
4-Chloroquinolin-8-amine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Chloroquinolin-8-amine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
81764-16-1
:
Formula:
C₉H₇ClN₂
MDL Number:
MFCD18448245
:
178.618 g/mol
Storage Temperature:
Ambient