Order Entry
Finland
ContactUsLinkComponent
6-Chloro-2-methyl-1-indanone 95%
6-Chloro-2-methyl-1-indanone 95%
Catalog # APOSOR916784-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Chloro-2-methyl-1-indanone 95%
Catalog # APOSOR916784-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.631 g/mol
Storage Temperature:Ambient
CAS Number:343852-44-8
Formula:C₁₀H₉ClO
MDL Number:MFCD16302118