4,5,6,7-Tetrahydro-1H-benzoimidazole 95%
Catalog # APOSOR912584-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:122.168 g/mol
Storage Temperature:Ambient
CAS Number:3752-24-7
Formula:C₇H₁₀N₂
MDL Number:MFCD00829762