Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-2-methoxy-4-methylaniline
5-Chloro-2-methoxy-4-methylaniline 96%
5-Chloro-2-methoxy-4-methylaniline 96%
5-Chloro-2-methoxy-4-methylaniline 96%
Supplier:
Apollo Scientific
Certificates
Expand image
5-Chloro-2-methoxy-4-methylaniline 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
569688-67-1
:
Formula:
C₈H₁₀ClNO
MDL Number:
MFCD14537235
:
171.624 g/mol
Storage Temperature:
Ambient