Order Entry
Finland
ContactUsLinkComponent
3-Chloro-1,3,4,5-tetrahydro-2H-1-benzazepin-2-one 95%
3-Chloro-1,3,4,5-tetrahydro-2H-1-benzazepin-2-one 95%
Catalog # APOSOR912146-5G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-1,3,4,5-tetrahydro-2H-1-benzazepin-2-one 95%
Catalog # APOSOR912146-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :195.646 g/mol
Storage Temperature:Ambient
CAS Number:86499-23-2
Formula:C₁₀H₁₀ClNO
MDL Number:MFCD09260932