Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-[(Diethylamino)methyl]benzonitrile
4-[(Diethylamino)methyl]benzonitrile 98%
4-[(Diethylamino)methyl]benzonitrile 98%
4-[(Diethylamino)methyl]benzonitrile 98%
Supplier:
Apollo Scientific
Certificates
Expand image
4-[(Diethylamino)methyl]benzonitrile 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
69293-74-9
:
Formula:
C₁₂H₁₆N₂
MDL Number:
MFCD09948026
:
188.269 g/mol
Storage Temperature:
Ambient