
4-(5-Chlorothien-2-yl)-1,3-thiazol-2-amine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:123971-45-9
:
Formula:C₇H₅ClN₂S₂
MDL Number:MFCD01312979
:216.711 g/mol
Melting Pt:129…130 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together