Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-8-methoxyquinoline
4-Bromo-8-methoxyquinoline
4-Bromo-8-methoxyquinoline
Irritant
4-Bromo-8-methoxyquinoline
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Bromo-8-methoxyquinoline
Supplier:
Apollo Scientific
About this item
CAS Number:
103028-31-5
:
Formula:
C₁₀H₈BrNO
MDL Number:
MFCD08063209
:
238.081 g/mol
Melting Pt:
97.2…97.4 °C
:
GHS07
Storage Temperature:
Ambient