Order Entry
Finland
ContactUsLinkComponent
4-(Bromomethyl)aniline, N-BOC protected 97%
Corrosive
Irritant
4-(Bromomethyl)aniline, N-BOC protected 97%
Catalog # APOSOR55614-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
4-(Bromomethyl)aniline, N-BOC protected 97%
Catalog # APOSOR55614-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :286.165 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
 :GHS07
CAS Number:239074-27-2
Formula:C₁₂H₁₆BrNO₂
MDL Number:MFCD11616718