Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 4-chloro-6-(trifluoromethyl)nicotinate
Ethyl 4-chloro-6-(trifluoromethyl)nicotinate 97%
Ethyl 4-chloro-6-(trifluoromethyl)nicotinate 97%
Ethyl 4-chloro-6-(trifluoromethyl)nicotinate 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Ethyl 4-chloro-6-(trifluoromethyl)nicotinate 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1196146-35-6
:
Formula:
C₉H₇ClF₃NO₂
MDL Number:
MFCD13189819
:
253.606 g/mol
Storage Temperature:
Refrigerator