Order Entry
Finland
ContactUsLinkComponent
N4-[4-[4-(N-[4-[2-(1-Naphthyl)anilino]phenyl]anilino)phenyl]phenyl]-N1-[2-(1-naphthyl)phenyl]-N4-phenyl-benzene-1,4-diamine 99%
N4-[4-[4-(N-[4-[2-(1-Naphthyl)anilino]phenyl]anilino)phenyl]phenyl]-N1-[2-(1-naphthyl)phenyl]-N4-phenyl-benzene-1,4-diamine 99%
Catalog # APOSOR51022-5G
Supplier:  Apollo Scientific
CAS Number:  
N4-[4-[4-(N-[4-[2-(1-Naphthyl)anilino]phenyl]anilino)phenyl]phenyl]-N1-[2-(1-naphthyl)phenyl]-N4-phenyl-benzene-1,4-diamine 99%
Catalog # APOSOR51022-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :923.151 g/mol
Storage Temperature:Ambient
CAS Number:209980-47-2
 :MFCD22581310
Formula:C₆₈H₅₀N₄
MDL Number:MFCD23135499