Order Entry
Finland
ContactUsLinkComponent
 
3-Methyl-1H-pyrrole-2-carbaldehyde ≥98%
Catalog # APOSOR510123-100MG
Supplier:  Apollo Scientific
CAS Number:  
3-Methyl-1H-pyrrole-2-carbaldehyde ≥98%
Catalog # APOSOR510123-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :109.126 g/mol
Storage Temperature:Ambient
CAS Number:24014-18-4
Formula:C₆H₇NO
MDL Number:MFCD00030343