Hydroquinidine 1,4-phthalazinediyl ether mixture
Catalog # APOSOR48099-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:778.98 g/mol
Storage Temperature:Ambient
CAS Number:148618-32-0
Formula:C₄₈H₅₄N₆O₄
MDL Number:MFCD00198107