Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Di-p-tolylethylenediamine
1,2-Di-p-tolylethylenediamine
1,2-Di-p-tolylethylenediamine
APOSOR471165-1G
:
Apollo Scientific
:
1,2-Di-p-tolylethylenediamine
APOSOR471165-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
240.343 g/mol
:
Ambient
:
124842-96-2
:
C₁₆H₂₀N₂
:
MFCD06658956