1-(2-Acetoxyethyl)pyrazole-4-boronic acid pinacol ester
Catalog # APOSOR470884-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:280.128 g/mol
Storage Temperature:Ambient
CAS Number:1251731-71-1
Formula:C₁₃H₂₁BN₂O₄
MDL Number:MFCD14707223