Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-3,6-dichloropyridine
2-Amino-3,6-dichloropyridine
2-Amino-3,6-dichloropyridine
2-Amino-3,6-dichloropyridine
Supplier:
Apollo Scientific
Certificates
Expand image
2-Amino-3,6-dichloropyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
313535-01-2
:
Formula:
C₅H₄Cl₂N₂
MDL Number:
MFCD00463634
:
163.005 g/mol
Storage Temperature:
Ambient