4-(3-Piperidyl)-1-butanol
Catalog # APOSOR470564-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:157.253 g/mol
Storage Temperature:Ambient
CAS Number:1021910-91-7
Formula:C₉H₁₉NO
MDL Number:MFCD23381096