Order Entry
Finland
ContactUsLinkComponent
6-Pivalamidopyridine-3-boronic acid pinacol ester
6-Pivalamidopyridine-3-boronic acid pinacol ester
Catalog # APOSOR470426-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Pivalamidopyridine-3-boronic acid pinacol ester
Catalog # APOSOR470426-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :304.192 g/mol
Storage Temperature:Ambient
CAS Number:1092119-20-4
Formula:C₁₆H₂₅BN₂O₃
MDL Number:MFCD13191341