3-Acetamido-4-methylpyridazine
Catalog # APOSOR470296-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:151.166 g/mol
Storage Temperature:Ambient
CAS Number:1314406-37-5
Formula:C₇H₉N₃O
MDL Number:MFCD19442780