4-Methyl-1H-pyrrole-2-carbaldehyde
Catalog # APOSOR46634-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:109.126 g/mol
Storage Temperature:Ambient
CAS Number:24014-19-5
Formula:C₆H₇NO
MDL Number:MFCD08235196