Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fmoc-N-methyl-DL-alanine
Fmoc-N-methyl-DL-alanine 97%
Fmoc-N-methyl-DL-alanine 97%
Fmoc-N-methyl-DL-alanine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Fmoc-N-methyl-DL-alanine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1362858-88-5
:
Formula:
C₁₉H₁₉NO₄
MDL Number:
MFCD18263130
:
325.358 g/mol
Storage Temperature:
Ambient