1-(3-Chlorophenyl)-cyclobutanemethanamine
Catalog # APOSOR451128-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:195.689 g/mol
Storage Temperature:Ambient
CAS Number:115816-34-7
Formula:C₁₁H₁₄ClN
MDL Number:MFCD09904043