Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Methoxyquinoline-3-carboxylic acid
6-Methoxyquinoline-3-carboxylic acid
6-Methoxyquinoline-3-carboxylic acid
6-Methoxyquinoline-3-carboxylic acid
Supplier:
Apollo Scientific
Certificates
Expand image
6-Methoxyquinoline-3-carboxylic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
71082-47-8
:
Formula:
C₁₁H₉NO₃
MDL Number:
MFCD08437579
:
203.194 g/mol
Melting Pt:
273 °C
Storage Temperature:
Ambient