
1-Ethyl-4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1047636-01-0
:
Formula:C₁₂H₂₁BN₂O₂
MDL Number:MFCD21099436
:236.118 g/mol
:GHS07
Storage Temperature:Refrigerator